under table shelf that is compatible with 24 x 24 inch optical breadboard and I-125 isolator frame. It clips onto the cross bracket of the Isostation frame and provides a large shelf depth, while still giving leg clearance. The surface provides 200 l 1 * 1 ST
Lieferant: DISCOVER ECHO
under table shelf that is compatible with 24 x 24 inch optical breadboard and I-125 isolator frame. It clips onto the cross bracket of the Isostation frame and provides a large shelf depth, while still giving leg clearance. The surface provides 200 l 1 * 1 ST