Wall Bracket for 30 L Bio-bin® Boxes
Lieferant: ECONIX LTD
EXBB30WB
ECONEXBB30WBEA 218 EUR
ECONEXBB30WB
Wall Bracket for 30 L Bio-bin® Boxes
Boxen Behälter zur Abfallentsorgung
This wall bracket is used in areas where the bin needs to be kept off the floor.
  • Easy to attach to a wall or to the side of a trolley
  • Chrome plated for easy cleaning
  • Fixed into place with M8 screws and plugs depending on the wall type (not supplied)
Bestellinformation: Supplied without screws.