Organic reference standard, Betamethasone, Ph.Eur., RS
Lieferant: EDQM
B1000000
EDQMB1000000EA 245 EUR
EDQMB1000000
Organic reference standard, Betamethasone, Ph.Eur., RS
Standards Organische Standards
  • In Monographs 0312,0354,0388, and 0811
  • Solubility in water: less than 0,063 g/l