Organic reference standard, Calcium pantothenate, Ph.Eur., RS
Lieferant: EDQM
C0400000
EDQMC0400000EA
245
EUR
EDQMC0400000
Organic reference standard, Calcium pantothenate, Ph.Eur., RS
Standards
Organische Standards
- Solubility in water at 25 °C: 350 g/l
- Pharmaceutical industry applications