Digoxin

Lieferant: EDQM

D1900000
EDQMD1900000EA 245 EUR
EDQMD1900000
Digoxin
Digoxin
Formel: C₄₁H₆₄O₁₄
Molecular Weight: 780.95 g/mol
MDL: MFCD00003674
CAS-Nummer: 20830-75-5
EINECS: 244-068-1
UN: 2811
ADR: 6.1 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR