Organic reference standard, Oxymetazoline hydrochloride, Ph.Eur., RS
Lieferant: EDQM
O0290000
EDQMO0290000EA 245 EUR
EDQMO0290000
Organic reference standard, Oxymetazoline hydrochloride, Ph.Eur., RS
Standards Organische Standards
  • In Monograph 0943
  • Solubility in water: 100 to 1000 g/l