Organic reference standard, Pentamidine diisetionate, Ph.Eur., RS
Lieferant: EDQM
P0405000
EDQMP0405000EA 245 EUR
EDQMP0405000
Organic reference standard, Pentamidine diisetionate, Ph.Eur., RS
Standards Organische Standards
  • In Monograph 1137 and 1436
  • Solubility in water: 100 to 1000 g/l