Organic reference standard, Piroxicam, Ph.Eur., RS
Lieferant: EDQM
P2130000
EDQMP2130000EA 245 EUR
EDQMP2130000
Organic reference standard, Piroxicam, Ph.Eur., RS
Standards Organische Standards
  • In Monograph 0944
  • Solubility in water: less than 0,1 g/l