Vitamin A acetat

Lieferant: EDQM

Synonyme: alle-trans-Retinylacetat

R0300000
EDQMR0300000EA 245 EUR
EDQMR0300000
Vitamin A acetat
alle-trans-Retinylacetat
Formel: C₂₂H₃₂O₂
Molecular Weight: 328,49 g/mol
MDL: MFCD00019413
CAS-Nummer: 127-47-9
EINECS: 204-844-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR