Organic reference standard, Simvastatin, Ph.Eur., RS
Lieferant: EDQM
S0650000
EDQMS0650000EA
245
EUR
EDQMS0650000
Organic reference standard, Simvastatin, Ph.Eur., RS
Standards
Organische Standards
- In Monograph 1563
- Soluble in/miscibility with ethyl acetate and methanol
- Pharmaceutical applications