Organic reference standard, Sisomicin sulfate, Ph.Eur., RS
Lieferant: EDQM
S0660000
EDQMS0660000EA 245 EUR
EDQMS0660000
Organic reference standard, Sisomicin sulfate, Ph.Eur., RS
Standards Organische Standards
  • In Monograph 0331 and 1351
  • Partial solubility in water
  • Pharmaceutical applications