DL-α-Tocopherolacetat

Lieferant: EDQM

Synonyme: (±)-α-Tocopherol acetate, all-rac-α-Tocopheryl acetate, Vitamine E acetate, DL-alpha-Tocopheryl acetate, 2,5,7,8-Tetramethyl-2-(4,8,12-trimethyltridecyl)chroman-6-yl acetate

T1600000
EDQMT1600000EA 245 EUR
EDQMT1600000
DL-α-Tocopherolacetat
DL-α-Tocopherolacetat
Formel: C₃₁H₅₂O₃
Molecular Weight: 472,75 g/mol
MDL: MFCD00072042
CAS-Nummer: 7695-91-2
EINECS: 231-710-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR