Organic reference standard, Triamcinolone, Ph.Eur., RS
Lieferant: EDQM
T1899990
EDQMT1899990EA 245 EUR
EDQMT1899990
Organic reference standard, Triamcinolone, Ph.Eur., RS
Standards Organische Standards
  • In Monograph 1376
  • Solubility in/miscible with DMSO and methanol