Organic reference standard, Perindopril for stereochemical purity, Ph.Eur., RS
Lieferant: EDQM
Y0000207
EDQMY0000207EA
245
EUR
EDQMY0000207
Organic reference standard, Perindopril for stereochemical purity, Ph.Eur., RS
Standards
Organische Standards
- In Monograph 2019
- Soluble in ethanol (96%), dimethylsulfoxide (DMSO), and water
- Pharmaceutical applications