Organic reference standard, Perindopril for stereochemical purity, Ph.Eur., RS
Lieferant: EDQM
Y0000207
EDQMY0000207EA 245 EUR
EDQMY0000207
Organic reference standard, Perindopril for stereochemical purity, Ph.Eur., RS
Standards Organische Standards
  • In Monograph 2019
  • Soluble in ethanol (96%), dimethylsulfoxide (DMSO), and water
  • Pharmaceutical applications