D(-)-N-Methylglucamin

Lieferant: EDQM

Synonyme: (-)-N-Methyl-D-glucamin, (-)-1-Methylamino-1-desoxy-D-glucitol, Meglumin, (-)-1-Desoxy-1-(methylamino)-D-glucitol

Y0001209
EDQMY0001209EA 245 EUR
EDQMY0001209
D(-)-N-Methylglucamin
D(-)-N-Methylglucamin
Formel: H₃CNHCH₂(CH(OH))₄CH₂OH
Molecular Weight: 195,22 g/mol
Schmelzpunkt: 128…131 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004707
CAS-Nummer: 6284-40-8
EINECS: 228-506-9
Merck Index: 12,06154

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR