Experimentation kits, blood typing
Lieferant: EDVOTEK
140
EDVO140EA 60.5 EUR
EDVO140
Experimentation kits, blood typing
Lehrmittel Forensik-Lehrmittel Forensische Kits
ABO and Rh typing of blood left at the scene of a crime can help to narrow down a list of suspects. In this experiment, students use agglutination to identify the blood group of unknown blood samples as a step to identify a criminal.
  • For 10 lab groups
  • Complete in 45 minutes
Also required: Microscope with 400x magnification.
Lieferumfang: Kit includes instructions, control ABO Rh simulated blood samples, unknown simulated blood samples, transfer pipettes, and microscope slides.