Transformation of E. coli with pGAL™
Lieferant: EDVOTEK
221
EDVO221EA 185 EUR
EDVO221
Transformation of E. coli with pGAL™
Lehrmittel Biologie-Lehrmittel Mikrobiologie-Lehrmittel
In this experiment, students see a blue colour change in transformed cells due to the switching on of a gene. The pGAL plasmid gives a blue colour dye due to the production of the β-galactosidase protein by the lacZ gene. IPTG is not required in this experiment since pGAL contains the complete lacZ gene.
  • For 10 lab groups
  • Complete in 50 minutes and grow overnight
Also required: 37 °C incubator, water bath.
Lieferumfang: Kit includes instructions, LyphoCells, plasmid DNA, buffer, media, ampicillin, X-Gal, ReadyPour agar, petri dishes, sterile pipettes, loops and microtubes.