Long flange with weld stub
Lieferant: EDWARDS HIGH VACUUM METTLER
C10514316
EDWAC10514316EA 0 EUR
EDWAC10514316
Long flange with weld stub
Klemmen Klemmen für den allgemeinen Gebrauch
This flange fittings made from stainless steel 316L are ideal for rugged corrosion resistance in semiconductor processing and offer excellent repeatability in high vacuum applications.
  • Quality control of elastomer specifications
  • Fit-and-forget dependability