Formaldehyd 1000 µg/ml water
Lieferant: EHRENSTORFER
|
Gefahr
|
Synonyme:
Methanal, Formalin, Formol, Methylaldehyd
A13909000WA
EHERA13909000WAEA
39
EUR
EHERA13909000WA
Formaldehyd 1000 µg/ml water
Formaldehyd
Formel:
CH₂O Molecular Weight: 30.03 g/mol Siedepunkt: -19.3 Schmelzpunkt: -92 Lagertemperatur: Kühlschrank |
MDL:
MFCD00003274 CAS-Nummer: 50-00-0 Merck Index: 13,04259 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...