Histamin Dihydrochlorid 100 µg/ml in Wasser, farblos, Flüssigkeit
Lieferant: EHRENSTORFER
|
Achtung
|
Synonyme:
Histamine 2HCl, 4-(2-Aminoethyl)-1H-imidazole dihydrochloride, 2-(1H-Imidazol-4-yl)ethan-1-amine dihydrochloride
A14213050WL100
EHERA14213050WL100EA
81
EUR
EHERA14213050WL100
Histamin Dihydrochlorid 100 µg/ml in Wasser, farblos, Flüssigkeit
Histamin Dihydrochlorid
Formel:
C₅H₁₁Cl₂N₃ Molecular Weight: 184,07 g/mol |
MDL:
MFCD00012703 CAS-Nummer: 56-92-8 UN: 1648 ADR: 3,II |
Spezifikation Testergebnisse
Accurate mass | 183.033 |
Shipping temperature | 20 °C |
Odour | Aromatic |
Stability | Stable under normal conditions |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...