Tulathromycin A
Lieferant: EHRENSTORFER
Synonyme:
Draxxin
A17895290AL100
EHERA17895290AL100EA
51
EUR
EHERA17895290AL100
Tulathromycin A
Tulathromycin A
Formel:
C₄₁H₇₉N₃O₁₂ Molecular Weight: 806,08 g/mol |
MDL:
MFCD09838804 CAS-Nummer: 217500-96-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...