Acepromazinmaleat
Lieferant: EHRENSTORFER
C10010300
EHERC10010300EA
178
EUR
EHERC10010300
Acepromazinmaleat
Acepromazinmaleat
Formel:
C₂₃H₂₆N₂O₅S Molecular Weight: 442,53 g/mol |
CAS-Nummer:
3598-37-6 EINECS: 222-748-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...