2-Acetamido-5-nitrothiazol
Lieferant: EHRENSTORFER
Synonyme:
N-(5-Nitro-2-thiazolyl)acetamide, N-(5-Nitrothiazol-2-yl)acetamide
C10012500
EHERC10012500EA
103
EUR
EHERC10012500
2-Acetamido-5-nitrothiazol
2-Acetamido-5-nitrothiazol
Formel:
C₅H₅N₃O₃S Molecular Weight: 187,18 g/mol |
MDL:
MFCD00022438 CAS-Nummer: 140-40-9 EINECS: 205-414-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...