Metanilgelb

Lieferant: EHRENSTORFER

Synonyme: Acid Yellow 36, 3-(4-Anilinophenylazo)benzenesulfonic acid sodium salt

C10029500
EHERC10029500EA 158 EUR
EHERC10029500
Metanilgelb
Metanilgelb
Formel: C₁₈H₁₄N₃NaO₃S
Molecular Weight: 375,38 g/mol
MDL: MFCD00007486
CAS-Nummer: 587-98-4
EINECS: 209-608-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR