Bis(2-ethylhexyl)adipat

Lieferant: EHRENSTORFER
Achtung

Synonyme: Di-(2-ethylhexyl) adipate, Bis(2-ethylhexyl) hexanedioate, Dioctyl Adipate, Diisooctyl adipate, Bis(2-ethylhexyl) adipate

C10046000
EHERC10046000EA 71 EUR
EHERC10046000
Bis(2-ethylhexyl)adipat
Bis(2-ethylhexyl)adipat
Formel: C₂₂H₄₂O₄
Molecular Weight: 370,57 g/mol
MDL: MFCD00009496
CAS-Nummer: 103-23-1
EINECS: 203-090-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR