Ribit
Lieferant: EHRENSTORFER
Synonyme:
Ribitol, 1,2,3,4,5-Pentanpentol, Adonitrol, (2R,3s,4S)-1,2,3,4,5-Pentanpentol, Ribit, Adonit, Pentitol
C10046500
EHERC10046500EA
61
EUR
EHERC10046500
Ribit
Adonitol
Formel:
HOH₂C(CH(OH))₃CH₂OH Molecular Weight: 152.15 g/mol Schmelzpunkt: 100…103 °C |
MDL:
MFCD00064291 CAS-Nummer: 488-81-3 EINECS: 207-685-7 Merck Index: 13,00168 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...