2-Chlor-4-nitrobenzamid

Lieferant: EHRENSTORFER

Synonyme: Aklomid

C10049000
EHERC10049000EA 130 EUR
EHERC10049000
2-Chlor-4-nitrobenzamid
2-Chlor-4-nitrobenzamid
Formel: C₇H₅ClN₂O₃
MDL: MFCD00017119
CAS-Nummer: 3011-89-0
EINECS: 221-143-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR