Alachlor

Lieferant: EHRENSTORFER

C10060000
EHERC10060000EA 80 EUR
EHERC10060000
Alachlor
Alachlor
Formel: C₁₄H₂₀ClNO₂
CAS-Nummer: 15972-60-8
EINECS: 240-110-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR