2-Aminoazotoluol

Lieferant: EHRENSTORFER

C10202000
EHERC10202000EA 65 EUR
EHERC10202000
2-Aminoazotoluol
2-Aminoazotoluol
Formel: C₁₄H₁₅N₃
Molecular Weight: 225,29 g/mol
MDL: MFCD00007733
CAS-Nummer: 97-56-3
EINECS: 202-591-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR