4-Amino-3-fluorphenol

Lieferant: EHRENSTORFER
Gefahr

Synonyme: 2-Fluoro-4-hydroxyaniline

C10202400
EHERC10202400EA 221 EUR
EHERC10202400
4-Amino-3-fluorphenol
4-Amino-3-fluorphenol
Formel: C₆H₆FNO
Molecular Weight: 127,12 g/mol
MDL: MFCD00077452
CAS-Nummer: 399-95-1
EINECS: 402-230-0
UN: 3077
ADR: 9,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR