tert-Amylmethylether

Lieferant: EHRENSTORFER
Gefahr

Synonyme: TAME, METHOXYPENTANE™, Methyl 2-methyl-2-butyl ether, 1,1-Dimethylpropyl methyl ether, Methyl tert-pentyl ether

C10246440
EHERC10246440EA 40 EUR
EHERC10246440
tert-Amylmethylether
tert-Amylmethylether
Formel: C₆H₁₄O
Molecular Weight: 102,17 g/mol
Siedepunkt: 85-86 °C (Lit.)
Dichte: 0,76…0.78 g/mL (25 °C) (Lit.)
Flammpunkt: -7 °C
MDL: MFCD00010186
CAS-Nummer: 994-05-8
EINECS: 213-611-4
UN: 3271
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR