4-(2-Methyl-2-butanyl)phenol

Lieferant: EHRENSTORFER

Synonyme: p-(1,1-Dimethylpropyl)phenol, 4-(2-Methyl-2-butyl)phenol, 4-tert-Pentylphenol

C10247000
EHERC10247000EA 40 EUR
EHERC10247000
4-(2-Methyl-2-butanyl)phenol
4-tert-Amylphenol
Formel: C₁₁H₁₆O
Molecular Weight: 164,25 g/mol
MDL: MFCD00002369
CAS-Nummer: 80-46-6
EINECS: 201-280-9
UN: 2430
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR