Anilin-[Ring-D5]
Lieferant: EHRENSTORFER
Synonyme:
Aniline-[ring-²H₅]
C10262600
EHERC10262600EA
106
EUR
EHERC10262600
Anilin-[Ring-D5]
Anilin-[Ring-D5]
Formel:
C₆H₂D₅N Molecular Weight: 98,09 g/mol Schmelzpunkt: –6,3 °C Dichte: 1,077 g/cm³ (20 °C) |
CAS-Nummer:
4165-61-1 EINECS: 224-015-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...