DL-Arginin-hydrochlorid, kristalliner Feststoff
Lieferant: EHRENSTORFER
|
Achtung
|
Synonyme:
DL-Arginine xhydrochloride, DL-Arginine Hydrochloride, DL-(+/-)-Arginine hydrochloride, DL-(±)-Arginine Hydrochloride, H-DL-Arg-OH.HCl, H-DL-Arg-OH·HCl, H-DL-(±)-Arg-OH·HCl, DL-2-Amino-5-guanidinopentanoic acid hydrochloride monohydrate
C10300200
EHERC10300200EA
44
EUR
EHERC10300200
DL-Arginin-hydrochlorid, kristalliner Feststoff
DL-Arginine hydrochloride
Formel:
C₆H₁₄N₄O₂.HCl Molecular Weight: 210,66 g/mol |
MDL:
MFCD00064549 CAS-Nummer: 32042-43-6 EINECS: 230-571-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...