Desethylatrazin

Lieferant: EHRENSTORFER

Synonyme: 6-Chloro-N-(isopropyl)-1,3,5-triazine-2,4-diamine

C10331000
EHERC10331000EA 93 EUR
EHERC10331000
Desethylatrazin
Desethylatrazin
Formel: C₈H₁₄ClN₅
Molecular Weight: 215.69 g/mol
CAS-Nummer: 6190-65-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR