Azocyclotin
Lieferant: EHRENSTORFER
C10400000
EHERC10400000EA
72
EUR
EHERC10400000
Azocyclotin
Azocyclotin
Formel:
C₂₀H₃₅N₃Sn |
MDL:
MFCD00055290 CAS-Nummer: 41083-11-8 EINECS: 255-209-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...