Viktoriareinblau BO
Lieferant: EHRENSTORFER
Synonyme:
Basic Blue 7, N-(4-((4-(Diethylamino)phenyl)(4-(ethylamino)naphthalen-1-yl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-ethylethanaminium chloride
C10424000
EHERC10424000EA
158
EUR
EHERC10424000
Viktoriareinblau BO
Viktoriareinblau BO
Formel:
C₃₃H₄₀ClN₃ Molecular Weight: 514,1438 g/mol |
MDL:
MFCD00011877 CAS-Nummer: 2390-60-5 EINECS: 219-232-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...