Safranin O

Lieferant: EHRENSTORFER

Synonyme: 3,7-Diamino-2,8-dimethyl-5-phenylphenaziniumchlorid, Basisrot 2

C10424960
EHERC10424960EA 46 EUR
EHERC10424960
Safranin O
Safranin O
Formel: C₂₀H₁₉ClN₄
Molecular Weight: 350.85 g/mol
Dichte: 1.447 g/cm³ (22 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011759
CAS-Nummer: 477-73-6
EINECS: 207-518-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR