1,2-Benzisothiazol-3(2H)-on

Lieferant: EHRENSTORFER

C10536600
EHERC10536600EA 87 EUR
EHERC10536600
1,2-Benzisothiazol-3(2H)-on
1,2-Benzisothiazol-3(2H)-on
Formel: C₇H₅NOS
MDL: MFCD00044001
CAS-Nummer: 2634-33-5
EINECS: 220-120-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR