2,4-Diamino-6-phenyl-1,3,5-triazin

Lieferant: EHRENSTORFER

Synonyme: 6-Phenyl-1,3,5-triazin-2,4-diamin, 6-Phenyl-1,3,5-triazin-2,4-diyldiamin, Benzoguanamin, 2,4-Diamino-6-phenyl-s-triazin

C10537450
EHERC10537450EA 60 EUR
EHERC10537450
2,4-Diamino-6-phenyl-1,3,5-triazin
2,4-Diamino-6-phenyl-1,3,5-triazine
Formel: C₉H₉N₅
Molecular Weight: 187,2 g/mol
MDL: MFCD00023187
CAS-Nummer: 91-76-9
EINECS: 202-095-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR