Binapacryl (2-sec-Butyl-4,6-dinitrophenyl-3-methylcrotonat)
Lieferant: EHRENSTORFER
C10590000
EHERC10590000EA
83
EUR
EHERC10590000
Binapacryl (2-sec-Butyl-4,6-dinitrophenyl-3-methylcrotonat)
Binapacryl (2-sec-Butyl-4,6-dinitrophenyl-3-methylcrotonat)
Formel:
C₁₅H₁₈N₂O₆ |
CAS-Nummer:
485-31-4 EINECS: 207-612-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...