4,4'-Bis(dimethylamino)diphenylmethan

Lieferant: EHRENSTORFER

Synonyme: 4,4'-Methylenebis(N,N-dimethylaniline), Bis[4-(dimethylamino)phenyl]methane

C10651900
EHERC10651900EA 45 EUR
EHERC10651900
4,4'-Bis(dimethylamino)diphenylmethan
4,4'-Bis(dimethylamino)diphenylmethan
Formel: C₁₇H₂₂N₂
Molecular Weight: 254,38 g/mol
MDL: MFCD00008317
CAS-Nummer: 101-61-1
EINECS: 202-959-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR