Bis(2-hydroxypropyl) ether

Lieferant: EHRENSTORFER

Synonyme: 1,1'-Oxybis(propan-2-ol)(mixture of isomers)

C10653600
EHERC10653600EA 69.5 EUR
EHERC10653600
Bis(2-hydroxypropyl) ether
1,1'-Oxybis(propan-2-ol)
Formel: C₆H₁₄O₃
Molecular Weight: 134,1736 g/mol
Lagertemperatur: Room Temperature
MDL: MFCD00004538
CAS-Nummer: 110-98-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR