4,4'-(1-Methylpropyliden)diphenol
Lieferant: EHRENSTORFER
Synonyme:
2,2-Bis(4-hydroxyphenyl)butan
C10655670
EHERC10655670EA
65
EUR
EHERC10655670
4,4'-(1-Methylpropyliden)diphenol
4,4'-(1-Methylpropyliden)diphenol
Formel:
C₁₆H₁₈O₂ Molecular Weight: 242,32 g/mol Siedepunkt: 412 °C (1013 hPa) Schmelzpunkt: 124…125 °C |
MDL:
MFCD00020179 CAS-Nummer: 77-40-7 EINECS: 201-025-1 Merck Index: 12,01339 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...