(±)-1-Butoxy-2-propanol
Lieferant: EHRENSTORFER
|
Achtung
|
Synonyme:
(±)-Propylenglycolmonobutylether, Lösungsmittel 35
C10900100
EHERC10900100EA
60
EUR
EHERC10900100
(±)-1-Butoxy-2-propanol
(±)-1-Butoxy-2-propanol
Formel:
C₇H₁₆O₂ Molecular Weight: 132,2 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00021904 CAS-Nummer: 5131-66-8 EINECS: 225-878-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...