2,4-Dimethyl-6-(2-methyl-2-propanyl)phenol
Lieferant: EHRENSTORFER
C10931188
EHERC10931188EA
89
EUR
EHERC10931188
2,4-Dimethyl-6-(2-methyl-2-propanyl)phenol
6-tert-Butyl-2,4-xylenol
Formel:
C₁₂H₁₈O |
MDL:
MFCD00002234 CAS-Nummer: 1879-09-0 EINECS: 217-533-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...