Butyrophenon
Lieferant: EHRENSTORFER
Synonyme:
Phenylpropylketon
C10931900
EHERC10931900EA
35
EUR
EHERC10931900
Butyrophenon
Butyrophenon
Formel:
C₆H₅C(O)(CH₂)₂CH₃ Molecular Weight: 148,2 g/mol Siedepunkt: 220 °C (1013 hPa) Dichte: 0,988 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009397 CAS-Nummer: 495-40-9 EINECS: 207-799-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...