Carbamazepin

Lieferant: EHRENSTORFER

Synonyme: 5H-Dibenzo[b,f]azepine-5-carboxamide

C10968500
EHERC10968500EA 44 EUR
EHERC10968500
Carbamazepin
Carbamazepin
Formel: C₁₅H₁₂N₂O
Molecular Weight: 236.27 g/mol
MDL: MFCD00005073
CAS-Nummer: 298-46-4
EINECS: 206-062-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR