Carbaryl (1-Naphthylmethylcarbamat)
Lieferant: EHRENSTORFER
C10980000
EHERC10980000EA
70
EUR
EHERC10980000
Carbaryl (1-Naphthylmethylcarbamat)
Carbaryl (1-Naphthylmethylcarbamat)
Formel:
C₁₂H₁₁NO₂ Molecular Weight: 201.22 g/mol |
CAS-Nummer:
63-25-2 EINECS: 200-555-0 UN: 2757 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...