L(-)-Carnitin

Lieferant: EHRENSTORFER

Synonyme: Levocarnitine, L-Carnitine, Vitamin BT, (R)-3-Hydroxy-4-(trimethylammonio)butanoate, R)-(-)-3-Hydroxy-4-(trimethylammonio)butyrate

C11045500
EHERC11045500EA 44 EUR
EHERC11045500
L(-)-Carnitin
L(-)-Carnitin
Formel: C₇H₁₅NO₃
Molecular Weight: 161,20 g/mol
MDL: MFCD00038747
CAS-Nummer: 541-15-1
EINECS: 208-768-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR